Assay Details
Target Gene Details
Entrez Gene ID: | 14460 |
Gene Name: | GATA binding protein 1 |
Gene Aliases: |
Gata-1, Gf-1, eryf1 |
Location: |
Chr.X:7959260-7976677 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gata1 | NM_008089.2 | NP_032115.1 | ||
| AK146915.1 | BAE27527.1 | |||
| BC052653.1 | AAH52653.1 | |||
| X15763.1 | CAA33769.1 |
More Information
Set Membership: |
|

Genomic Map