Assay Details
Target Gene Details
Entrez Gene ID: | 245469 |
Gene Name: | PDZ domain containing 4 |
Gene Aliases: |
B230341P03, D130075J17Rik, Kiaa1444-hp, LU1, PDZRN4L, Pdzk4, Pdzrnk4l, Xlu |
Location: |
Chr.X:73793357-73824969 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pdzd4 | NM_001029868.2 | NP_001025039.1 | ||
| NM_001290540.1 | NP_001277469.1 | |||
| AK046101.1 | ||||
| BC056462.1 | AAH56462.1 |

Genomic Map