Assay Details
Target Gene Details
Entrez Gene ID: | 18630 |
Gene Name: | plasmacytoma expressed transcript 2 |
Gene Aliases: |
PC231, PC326, Pex3 |
Location: |
Chr.X:89403848-89409689 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pet2 | NM_008821.2 | 2 | 200 | NP_032847.2 |
| BC138080.1 | ||||
| M95564.1 | 2 | 197 | AAA39895.1 |
More Information
Set Membership: |
|

Genomic Map