Assay Details
Target Gene Details
Entrez Gene ID: | 18823 |
Gene Name: | proteolipid protein (myelin) 1 |
Gene Aliases: |
DM20, Plp, jimpy, jp, msd, rsh |
Location: |
Chr.X:136822746-136838582 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Plp1 | NM_001290561.1 | NP_001277490.1 | ||
| NM_001290562.1 | NP_001277491.1 | |||
| NM_011123.3 | NP_035253.1 | |||
| AK077564.1 | ||||
| AK134554.1 | BAE22184.1 | |||
| AK160872.1 | BAE36061.1 | |||
| AK197253.1 | ||||
| AK213737.1 | ||||
| AK215067.1 | ||||
| BC027010.1 | AAH27010.1 | |||
| M14674.1 | AAA39955.1 | |||
| M15442.1 | AAA39951.1 | |||
| M16472.1 | AAA39950.1 | |||
| M20752.1 | AAA39956.1 |
More Information
Set Membership: |
|

Genomic Map