Assay Details
Target Gene Details
Entrez Gene ID: | 18194 |
Gene Name: | NAD(P) dependent steroid dehydrogenase-like |
Gene Aliases: |
AI747449, Bpa, H105E3, Str, XAP104 |
Location: |
Chr.X:72918521-72958528 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nsdhl | NM_010941.3 | 3 | 360 | NP_035071.3 |
| AF100198.1 | 3 | 360 | AAD38448.1 | |
| AK013945.1 | 3 | 243 | ||
| AK088026.1 | 3 | 243 | ||
| AK140930.1 | 3 | 241 | BAE24522.1 | |
| BC019945.1 | 3 | 229 | AAH19945.1 | |
| BC052834.1 | 3 | 321 | AAH52834.1 | |
| KF356400.1 | 2 | 145 | ||
| KF356401.1 | 2 | 145 |
More Information
Set Membership: |
|

Genomic Map