Assay Details
Target Gene Details
Entrez Gene ID: | 18846 |
Gene Name: | plexin A3 |
Gene Aliases: |
PlexA3, Plxa3, Plxn3, Plxn4, SEX, mKIAA4078 |
Location: |
Chr.X:74329066-74344689 on Build GRCm38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Plxna3 | NM_008883.2 | 17 | 3200 | NP_032909.2 |
| AK220285.1 | 14 | 2059 | BAD90210.1 | |
| BC093482.1 | 17 | 3183 | ||
| D86950.1 | 17 | 3166 | BAA13190.1 |

Genomic Map