Assay Details
Target Gene Details
Entrez Gene ID: | 53381 |
Gene Name: | peroxiredoxin 4 |
Gene Aliases: |
AOE372, Prx-iv, Prx4, TRANK |
Location: |
Chr.X:155323918-155340749 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Prdx4 | NM_016764.4 | NP_058044.1 | ||
| AK005031.1 | BAB23758.1 | |||
| AK015920.1 | ||||
| AK146402.1 | BAE27143.1 | |||
| AK152255.1 | BAE31074.1 | |||
| BC003349.1 | AAH03349.1 | |||
| BC019578.1 | AAH19578.1 | |||
| U96746.1 | AAB57846.1 |
More Information
Set Membership: |
|

Genomic Map