Assay Details
Target Gene Details
Entrez Gene ID: | 54645 |
Gene Name: | GRIP1 associated protein 1 |
Gene Aliases: |
AI854681, DXImx47e, GRASP-1, Sfc10, mKIAA1167 |
Location: |
Chr.X:7789961-7820567 on Build GRCm38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gripap1 | NM_001290455.1 | NP_001277384.1 | ||
| NM_207670.2 | NP_997553.1 | |||
| AK156165.1 | BAE33608.1 | |||
| AK158098.1 | BAE34355.1 | |||
| AK172590.1 | BAE43083.1 | |||
| AK172623.1 | BAE43101.1 | |||
| AK172699.1 | BAE43133.1 | |||
| AW342118.1 | ||||
| BC138523.1 | ||||
| BC145310.1 | ||||
| X97571.1 | CAA66184.1 |
More Information
Set Membership: |
|

Genomic Map