Assay Details
Target Gene Details
Entrez Gene ID: | 68041 |
Gene Name: | Mid1 interacting protein 1 (gastrulation specific G12-like (zebrafish)) |
Gene Aliases: |
3110038L01Rik, Mig12, S14R |
Location: |
Chr.X:10717365-10719702 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mid1ip1 | NM_001166635.1 | 1 | 149 | NP_001160107.1 |
| NM_026524.4 | 1 | 149 | NP_080800.1 | |
| AK090003.1 | BAC41039.1 | |||
| AK151153.1 | BAE30159.1 | |||
| AK159166.1 | BAE34868.1 | |||
| AK160062.1 | 1 | 77 | BAE35598.1 | |
| AK172143.1 | BAE42845.1 | |||
| BC004014.1 | AAH04014.1 | |||
| BC052899.1 | AAH52899.1 |

Genomic Map