Assay Details
Target Gene Details
Entrez Gene ID: | 18846 |
Gene Name: | plexin A3 |
Gene Aliases: |
PlexA3, Plxa3, Plxn3, Plxn4, SEX, mKIAA4078 |
Location: |
Chr.X:74329066-74344689 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Plxna3 | NM_008883.2 | NP_032909.2 | ||
| AK220285.1 | BAD90210.1 | |||
| BC093482.1 | ||||
| D86950.1 | BAA13190.1 |

Genomic Map