Assay Details
Target Gene Details
Entrez Gene ID: | 171170 |
Gene Name: | muscleblind-like 3 (Drosophila) |
Gene Aliases: |
A530038J18Rik, AI661274, CHCR, E430034C16Rik, MBLX, MBLX39, MBXL |
Location: |
Chr.X:51113494-51206035 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mbnl3 | NM_134163.4 | NP_598924.1 | ||
| AK040522.1 | BAC30616.1 | |||
| AK088971.1 | BAC40678.1 | |||
| BC057157.1 | 1 | 191 | AAH57157.1 | |
| BY011002.1 |
More Information
Set Membership: |
|

Genomic Map