Assay Details
Target Gene Details
Entrez Gene ID: | 171170 |
Gene Name: | muscleblind-like 3 (Drosophila) |
Gene Aliases: |
A530038J18Rik, AI661274, CHCR, E430034C16Rik, MBLX, MBLX39, MBXL |
Location: |
Chr.X:51113494-51206035 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mbnl3 | NM_134163.4 | 4 | 610 | NP_598924.1 |
| AB077700.1 | 3 | 516 | BAB85650.1 | |
| AF466292.1 | 3 | 516 | AAL87669.1 | |
| AJ427920.1 | 3 | 516 | CAD20871.1 | |
| AK040522.1 | 3 | 363 | BAC30616.1 | |
| AK050908.1 | 3 | 329 | BAC34454.1 | |
| AK088971.1 | 4 | 623 | BAC40678.1 | |
| AK167342.1 | 3 | 329 | BAE39442.1 | |
| BC057157.1 | 4 | 848 | AAH57157.1 |
More Information
Set Membership: |
|

Genomic Map