Assay Details
Target Gene Details
Entrez Gene ID: | 16151 |
Gene Name: | inhibitor of kappaB kinase gamma |
Gene Aliases: |
1110037D23Rik, AI848108, AI851264, AW124339, IKK[g], NEMO |
Location: |
Chr.X:74393291-74456792 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ikbkg | NM_001136067.2 | 3 | 395 | NP_001129539.1 |
| NM_001161421.1 | 3 | 395 | NP_001154893.1 | |
| NM_001161422.1 | 2 | 213 | NP_001154894.1 | |
| NM_001161423.1 | 2 | 265 | NP_001154895.1 | |
| NM_001161424.1 | 2 | 265 | NP_001154896.1 | |
| NM_010547.2 | 2 | 291 | NP_034677.2 | |
| NM_178590.4 | 2 | 291 | NP_848705.1 | |
| AF069542.1 | 1 | 150 | AAC40153.1 | |
| AF513109.1 | 3 | 306 | AAP47160.1 | |
| AK037020.1 | 2 | 270 | BAC29672.1 | |
| AK148163.1 | 2 | 249 | BAE28385.1 | |
| AK154095.1 | 2 | 277 | BAE32372.1 | |
| AY112937.1 | 3 | 395 | AAM54725.1 | |
| BC021431.1 | 3 | 385 | AAH21431.1 | |
| BY768708.1 | 2 | 301 | ||
| CJ099503.1 | 3 | 380 | ||
| CN701499.1 | 2 | 213 | ||
| CN717929.1 | 2 | 206 | ||
| CV556906.1 | 2 | 231 |

Genomic Map