Assay Details
Target Gene Details
Entrez Gene ID: | 16151 |
Gene Name: | inhibitor of kappaB kinase gamma |
Gene Aliases: |
1110037D23Rik, AI848108, AI851264, AW124339, IKK[g], NEMO |
Location: |
Chr.X:74393291-74456792 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ikbkg | NM_001136067.2 | NP_001129539.1 | ||
| NM_001161421.1 | NP_001154893.1 | |||
| NM_001161422.1 | NP_001154894.1 | |||
| NM_001161423.1 | NP_001154895.1 | |||
| NM_001161424.1 | NP_001154896.1 | |||
| NM_010547.2 | NP_034677.2 | |||
| NM_178590.4 | NP_848705.1 | |||
| AF069542.1 | AAC40153.1 | |||
| AF513109.1 | AAP47160.1 | |||
| AK037020.1 | BAC29672.1 | |||
| AK148163.1 | BAE28385.1 | |||
| AK154095.1 | BAE32372.1 | |||
| AK188269.1 | ||||
| AY112937.1 | AAM54725.1 | |||
| BC021431.1 | AAH21431.1 | |||
| CV556906.1 |

Genomic Map