Assay Details
Target Gene Details
Entrez Gene ID: | 19703 |
Gene Name: | renin binding protein |
Gene Aliases: |
Age, Rnbp |
Location: |
Chr.X:73922121-73930850 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Renbp | NM_001164704.1 | NP_001158176.1 | ||
| NM_023132.3 | NP_075621.3 | |||
| AK014347.1 | BAB29288.1 | |||
| AK146652.1 | BAE27332.1 | |||
| AK167648.1 | BAE39699.1 | |||
| AK182381.1 | ||||
| AK186123.1 | ||||
| AK194143.1 | ||||
| AK200509.1 | ||||
| AK205945.1 | ||||
| AK212491.1 | ||||
| AK215192.1 | ||||
| AK216097.1 | ||||
| AY183138.1 | AAO66340.1 | |||
| BC014821.1 | AAH14821.1 |
More Information
Set Membership: |
|

Genomic Map