Assay Details
Target Gene Details
Entrez Gene ID: | 56774 |
Gene Name: | solute carrier family 6 (neurotransmitter transporter), member 14 |
Gene Aliases: |
1110007A17Rik, 9030613J17Rik, ATB0plus, CATB0plus |
Location: |
Chr.X:21714900-21742358 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc6a14 | NM_020049.4 | 4 | 717 | NP_064433.3 |
| AB033285.1 | 4 | 475 | BAA94300.1 | |
| AF161714.1 | 4 | 717 | AAD49320.1 | |
| AF320226.1 | 4 | 661 | AAK43541.1 | |
| AK018553.1 | 4 | 576 | BAB31272.1 | |
| BC129908.1 | 4 | 508 | ||
| BC129909.1 | 4 | 508 |
More Information
Set Membership: |
|

Genomic Map