Assay Details
Target Gene Details
Entrez Gene ID: | 26896 |
Gene Name: | mediator complex subunit 14 |
Gene Aliases: |
9930001L01Rik, AU041628, Crsp2, ENSMUSG00000073278, Gm641, ORF1, Trap170 |
Location: |
Chr.X:12675368-12762042 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Med14 | NM_001048208.1 | NP_001041673.1 | ||
| NM_012005.3 | NP_036135.3 | |||
| AB019028.1 | BAA76610.1 | |||
| AK036758.1 | ||||
| AK084044.1 | ||||
| AK163306.1 | BAE37289.1 |
More Information
Set Membership: |
|

Genomic Map