Assay Details
Target Gene Details
Entrez Gene ID: | 74469 |
Gene Name: | TAF7-like RNA polymerase II, TATA box binding protein (TBP)-associated factor |
Gene Aliases: |
4933438I11Rik, 50kDa, Taf2q |
Location: |
Chr.X:134460116-134476526 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Taf7l | NM_028958.4 | NP_083234.2 | ||
| AF285574.1 | AAK31953.1 | |||
| AK017109.1 | BAB30600.1 | |||
| BC106854.2 | AAI06855.1 |
More Information
Set Membership: |
|

Genomic Map