Assay Details
Target Gene Details
Entrez Gene ID: | 22201 |
Gene Name: | ubiquitin-like modifier activating enzyme 1 |
Gene Aliases: |
A1S9, Sbx, Ube-1, Ube1x |
Location: |
Chr.X:20658302-20683179 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Uba1 | NM_001276316.1 | 1 | 89 | NP_001263245.1 |
| NM_009457.4 | 1 | 89 | NP_033483.2 | |
| AK088057.1 | BAC40121.1 | |||
| AK088528.1 | BAC40405.1 | |||
| BC138200.1 | ||||
| BC145984.1 | ||||
| BY004827.1 | 1 | 91 | ||
| D10576.1 | BAA01433.1 |
More Information
Set Membership: |
|

Genomic Map