Assay Details
Target Gene Details
Entrez Gene ID: | 22201 |
Gene Name: | ubiquitin-like modifier activating enzyme 1 |
Gene Aliases: |
A1S9, Sbx, Ube-1, Ube1x |
Location: |
Chr.X:20658302-20683179 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Uba1 | NM_001136085.2 | 11 | 1369 | NP_001129557.1 |
| NM_001276316.1 | 12 | 1461 | NP_001263245.1 | |
| NM_001276317.1 | 11 | 1364 | NP_001263246.1 | |
| NM_009457.4 | 11 | 1364 | NP_033483.2 | |
| AK081808.1 | 11 | 1335 | ||
| AK088057.1 | 11 | 1246 | BAC40121.1 | |
| AK088528.1 | 11 | 1246 | BAC40405.1 | |
| AK143416.1 | 11 | 1270 | BAE25369.1 | |
| AK171667.1 | 11 | 1335 | BAE42599.1 | |
| AK185680.1 | 2 | 204 | ||
| AK195016.1 | 2 | 192 | ||
| AK205753.1 | ||||
| AK206372.1 | 2 | 140 | ||
| AK206443.1 | 2 | 136 | ||
| AK209084.1 | 2 | 180 | ||
| AK210744.1 | 2 | 193 | ||
| BC058630.1 | 11 | 1310 | AAH58630.1 | |
| BC138200.1 | 11 | 1227 | ||
| BC145984.1 | 11 | 1228 | ||
| D10576.1 | 11 | 1230 | BAA01433.1 |
More Information
Set Membership: |
|

Genomic Map