Assay Details
Target Gene Details
Entrez Gene ID: | 22201 |
Gene Name: | ubiquitin-like modifier activating enzyme 1 |
Gene Aliases: |
A1S9, Sbx, Ube-1, Ube1x |
Location: |
Chr.X:20658302-20683179 on Build GRCm38 |
Assay Gene Location: | Within Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Uba1 | NM_001136085.2 | 20 | 2643 | NP_001129557.1 |
| NM_001276316.1 | 21 | 2735 | NP_001263245.1 | |
| NM_001276317.1 | 20 | 2638 | NP_001263246.1 | |
| NM_009457.4 | 20 | 2638 | NP_033483.2 | |
| AK081808.1 | 20 | 2612 | ||
| AK088057.1 | 20 | 2520 | BAC40121.1 | |
| AK088528.1 | 20 | 2520 | BAC40405.1 | |
| AK143416.1 | 20 | 2544 | BAE25369.1 | |
| AK171667.1 | 20 | 2609 | BAE42599.1 | |
| BC058630.1 | 20 | 2584 | AAH58630.1 | |
| BC138200.1 | 20 | 2501 | ||
| BC145984.1 | 20 | 2502 | ||
| D10576.1 | 20 | 2504 | BAA01433.1 | |
| X62580.1 | 5 | 573 | CAA44465.1 |
More Information
Set Membership: |
|

Genomic Map