Assay Details
Target Gene Details
Entrez Gene ID: | 22201 |
Gene Name: | ubiquitin-like modifier activating enzyme 1 |
Gene Aliases: |
A1S9, Sbx, Ube-1, Ube1x |
Location: |
Chr.X:20658302-20683179 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Uba1 | NM_001136085.2 | NP_001129557.1 | ||
| NM_001276316.1 | NP_001263245.1 | |||
| NM_001276317.1 | NP_001263246.1 | |||
| NM_009457.4 | NP_033483.2 | |||
| AK081808.1 | ||||
| AK088057.1 | BAC40121.1 | |||
| AK088528.1 | BAC40405.1 | |||
| AK143416.1 | BAE25369.1 | |||
| AK171667.1 | BAE42599.1 | |||
| AK177712.1 | ||||
| AK178664.1 | ||||
| AK180642.1 | ||||
| AK185591.1 | ||||
| AK186400.1 | ||||
| AK186677.1 | ||||
| AK188893.1 | ||||
| AK194086.1 | ||||
| AK210112.1 | ||||
| BC058630.1 | AAH58630.1 | |||
| BC138200.1 | ||||
| BC145984.1 | ||||
| D10576.1 | BAA01433.1 |
More Information
Set Membership: |
|

Genomic Map