Assay Details
Target Gene Details
Entrez Gene ID: | 236733 |
Gene Name: | ubiquitin specific peptidase 11 |
Gene Aliases: |
6230415D12Rik, mKIAA4085 |
Location: |
Chr.X:20703909-20720539 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 12 - Intron 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Usp11 | NM_145628.4 | NP_663603.3 | ||
| AK031767.1 | BAC27541.1 | |||
| AK049053.1 | BAC33526.1 | |||
| AK169116.1 | BAE40896.1 | |||
| AK220478.1 | BAD90502.1 | |||
| BC005470.1 | AAH05470.1 |
More Information
Set Membership: |
|

Genomic Map