Assay Details
Target Gene Details
Entrez Gene ID: | 107868 |
Gene Name: | ubiquitin specific peptidase 9, Y chromosome |
Gene Aliases: |
Dffry, Fafl2 |
Location: |
Chr.Y:1298961-1459782 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 37 - Exon 38 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Usp9y | NM_148943.2 | NP_683745.2 | ||
| AJ307017.1 | CAC38831.1 |
More Information
Set Membership: |
|

Genomic Map