Assay Details
Target Gene Details
Entrez Gene ID: | 69821 |
Gene Name: | mitochondrial transcription termination factor 4 |
Gene Aliases: |
1810059A23Rik, 4933412H03Rik, Mterfd2 |
Location: |
Chr.1:93299211-93305965 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mterf4 | NM_178051.3 | 4 | 2027 | NP_835152.1 |
| AK079098.1 | 4 | 1808 |
Target Gene Details
Entrez Gene ID: | 208777 |
Gene Name: | sushi, nidogen and EGF-like domains 1 |
Gene Aliases: |
6720455I24Rik, AI197264, AI642697, D430044C15Rik, SST3, Snep |
Location: |
Chr.1:93235815-93301065 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sned1 | AJ584850.1 | 31 | 8581 | CAE48492.1 |

Genomic Map