Assay Details
Target Gene Details
Entrez Gene ID: | 51801 |
Gene Name: | receptor (calcitonin) activity modifying protein 1 |
Gene Aliases: |
9130218E19Rik |
Location: |
Chr.1:91179822-91225196 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ramp1 | NM_001168392.1 | 4 | 672 | NP_001161864.1 |
| NM_016894.3 | NP_058590.1 | |||
| NM_178401.3 | 3 | 593 | NP_848488.2 | |
| AB019046.1 | BAA76617.1 | |||
| AF146522.1 | AAD35018.1 | |||
| AF209904.1 | AAF21036.1 | |||
| AJ250489.1 | CAB59511.1 | |||
| AK144331.1 | 3 | 593 | BAE25837.1 | |
| AK157896.1 | BAE34250.1 | |||
| AK165238.1 | BAE38095.1 | |||
| BC012644.1 | AAH12644.1 | |||
| BY707433.1 | 4 | 453 | ||
| BY733497.1 | 3 | 593 | ||
| CB522711.1 | 2 | 230 |
More Information
Set Membership: |
|

Genomic Map