Assay Details
Target Gene Details
Entrez Gene ID: | 240913 |
Gene Name: | a disintegrin-like and metallopeptidase (reprolysin type) with thrombospondin type 1 motif, 4 |
Gene Aliases: |
3830423K05, ADAM-TS4, ADAMTS-2, ADMP-1, mKIAA0688 |
Location: |
Chr.1:171250422-171259922 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Adamts4 | NM_172845.2 | 1 | 347 | NP_766433.1 |
| AK083534.1 | 1 | 347 | BAC38944.1 | |
| AK164900.1 | 1 | 343 | BAE37959.1 | |
| BC027773.1 | AAH27773.1 |
More Information
Set Membership: |
|

Genomic Map