Assay Details
Target Gene Details
Entrez Gene ID: | 271639 |
Gene Name: | adenylate cyclase 10 |
Gene Aliases: |
4930431D04Rik, 4931412F17, Sacy, sAC |
Location: |
Chr.1:165483869-165576778 on Build GRCm38 |
Assay Gene Location: | Within Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Adcy10 | NM_173029.3 | 20 | 2882 | NP_766617.2 |
| AK029849.1 | 20 | 2884 | BAC26642.1 | |
| AK133181.1 | 19 | 2770 | BAE21546.1 | |
| BC138448.1 | 20 | 2763 |

Genomic Map