Assay Details
Target Gene Details
Entrez Gene ID: | 13849 |
Gene Name: | epoxide hydrolase 1, microsomal |
Gene Aliases: |
AI195553, Eph-1, Eph1, Epxh1, mEH |
Location: |
Chr.1:180989556-181017495 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ephx1 | NM_010145.2 | 9 | 1312 | NP_034275.1 |
| AK018249.1 | 9 | 1321 | BAB31132.1 | |
| AK194514.1 | ||||
| AK204696.1 | ||||
| BC029636.1 | 9 | 1311 | AAH29636.1 | |
| BC045194.1 | 9 | 1312 | AAH45194.1 | |
| BC057857.1 | 9 | 1265 | AAH57857.1 | |
| BC061493.1 | 9 | 1338 | AAH61493.1 | |
| U89491.1 | 9 | 1281 | AAB49762.1 |
More Information
Set Membership: |
|

Genomic Map