Assay Details
Target Gene Details
Entrez Gene ID: | 433323 |
Gene Name: | sphingosine-1-phosphate phosphotase 2 |
Gene Aliases: |
SPPase2, Spp2 |
Location: |
Chr.1:78309501-78420289 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sgpp2 | NM_001004173.2 | NP_001004173.1 | ||
| AK087798.1 | 4 | 1093 | ||
| BC049995.1 | AAH49995.1 | |||
| BC147843.1 | ||||
| BC147850.1 | ||||
| BY234834.1 | ||||
| BY297240.1 |
More Information
Set Membership: |
|

Genomic Map