Assay Details
Target Gene Details
Entrez Gene ID: | 18582 |
Gene Name: | phosphodiesterase 6D, cGMP-specific, rod, delta |
Gene Aliases: |
AI841218, PrBP/delta |
Location: |
Chr.1:86543013-86582793 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pde6d | NM_008801.2 | 5 | 711 | NP_032827.1 |
| AF039216.1 | 5 | 670 | AAC25686.1 | |
| AK141209.1 | 5 | 717 | BAE24588.1 | |
| AK165701.1 | 5 | 936 | ||
| AK170067.1 | 5 | 723 | BAE41543.1 | |
| AK188050.1 | 1 | 75 | ||
| BC005636.1 | 5 | 711 | AAH05636.1 |
More Information
Set Membership: |
|

Genomic Map