Assay Details
Target Gene Details
Entrez Gene ID: | 67556 |
Gene Name: | phosphatidylinositol glycan anchor biosynthesis, class M |
Gene Aliases: |
4933437L05Rik, AI644422, AV302080, C920011G20Rik |
Location: |
Chr.1:172376531-172384099 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pigm | NM_026234.4 | 1 | 1261 | NP_080510.1 |
| AK018560.1 | 1 | 1214 | BAB31275.1 | |
| AK040397.1 | 1 | 1237 | BAC30585.1 | |
| AK043216.1 | 1 | 165 | BAC31497.1 | |
| AK044641.1 | 1 | 1230 | ||
| AK080242.1 | 1 | 1222 | BAC37857.1 | |
| AK082746.1 | 1 | 1259 | ||
| AK088117.1 | 1 | 1241 | BAC40156.1 | |
| AK148197.1 | 1 | 1197 | ||
| BC005650.1 | 1 | 1246 | AAH05650.1 | |
| BC083115.1 | 1 | 1237 | AAH83115.1 |
More Information
Set Membership: |
|

Genomic Map