Assay Details
Target Gene Details
Entrez Gene ID: | 215303 |
Gene Name: | calcium/calmodulin-dependent protein kinase I gamma |
Gene Aliases: |
CLICK-III, CaMKIgamma |
Location: |
Chr.1:193346346-193370292 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Camk1g | NM_144817.2 | 11 | 1127 | NP_659066.1 |
| AF428262.1 | 11 | 1075 | AAL28101.1 | |
| AY212936.1 | 10 | 1007 | AAP29965.1 | |
| BC021840.1 | 11 | 1076 | AAH21840.1 |
More Information
Set Membership: |
|

Genomic Map