Assay Details
Target Gene Details
Entrez Gene ID: | 12334 |
Gene Name: | calpain 2 |
Gene Aliases: |
AI326419, Capa-2, Capa2, m-calpain, m-calpin |
Location: |
Chr.1:182467259-182517483 on Build GRCm38 |
Assay Gene Location: | Within Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Capn2 | NM_009794.3 | 21 | 2534 | NP_033924.2 |
| AF015038.1 | 21 | 2480 | AAB94029.1 | |
| AK052141.1 | 19 | 2123 | BAC34855.1 | |
| AK150695.1 | 21 | 2508 | BAE29774.1 | |
| AK152094.1 | 21 | 2508 | BAE30942.1 | |
| AK152790.1 | 21 | 2511 | ||
| AK152994.1 | 21 | 2511 | ||
| AK154855.1 | 21 | 2548 | BAE32880.1 | |
| AK178489.1 | ||||
| AK189547.1 | ||||
| AK212305.1 | ||||
| AK216791.1 | ||||
| AK219579.1 | 1 | 261 | ||
| BC054726.1 | 21 | 2534 | AAH54726.1 | |
| D38117.1 | 21 | 2575 | BAA22964.1 |
More Information
Set Membership: |
|

Genomic Map