Assay Details
Target Gene Details
Entrez Gene ID: | 226413 |
Gene Name: | lactase |
Gene Aliases: |
Gm100, LAC, LPH, Lphl |
Location: |
Chr.1:128284764-128328318 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lct | NM_001081078.1 | 7 | 2166 | NP_001074547.1 |
| AK158042.1 | 1 | 268 | BAE34332.1 |
More Information
Set Membership: |
|

Genomic Map