Assay Details
Target Gene Details
Entrez Gene ID: | 240752 |
Gene Name: | phosphoinositide-3-kinase, class 2, beta polypeptide |
Gene Aliases: |
C330011J12Rik, Gm199, PI3K-C2beta, PI3KC2beta |
Location: |
Chr.1:133045672-133108688 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pik3c2b | NM_001099276.2 | 2 | 656 | NP_001092746.2 |
| AK049181.1 | 2 | 629 | ||
| AK165803.1 | 2 | 657 | ||
| BC132141.1 | 2 | 304 |
More Information
Set Membership: |
|

Genomic Map