Assay Details
Target Gene Details
Entrez Gene ID: | 12778 |
Gene Name: | atypical chemokine receptor 3 |
Gene Aliases: |
AW541270, CXC-R7, CXCR-7, Cmkor1, Cxcr7, RDC-1, Rdc1 |
Location: |
Chr.1:90203980-90215724 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ackr3 | NM_001271607.1 | 3 | 1220 | NP_001258536.1 |
| NM_007722.4 | 2 | 937 | NP_031748.2 | |
| AF000236.1 | 2 | 891 | AAB71343.1 | |
| BC015254.1 | 2 | 852 | AAH15254.1 |

Genomic Map