Assay Details
Target Gene Details
Entrez Gene ID: | 67997 |
Gene Name: | DEAD (Asp-Glu-Ala-Asp) box polypeptide 59 |
Gene Aliases: |
1210002B07Rik, 4833411G06Rik |
Location: |
Chr.1:136415271-136456190 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ddx59 | NM_026500.3 | 5 | 1397 | NP_080776.1 |
| AK004833.1 | 5 | 1398 | BAB23601.1 | |
| AK133731.1 | 5 | 1359 | BAE21808.1 | |
| BC023840.1 | 5 | 1324 | AAH23840.1 |

Genomic Map