Assay Details
Target Gene Details
Entrez Gene ID: | 22779 |
Gene Name: | IKAROS family zinc finger 2 |
Gene Aliases: |
A730095J18Rik, Helios, Zfpn1a2, Znfn1a2 |
Location: |
Chr.1:69531207-69687351 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ikzf2 | NM_011770.4 | 8 | 2524 | NP_035900.2 |
| AK047925.1 | 8 | 2386 | BAC33191.1 |
More Information
Set Membership: |
|

Genomic Map