Assay Details
Target Gene Details
Entrez Gene ID: | 257630 |
Gene Name: | interleukin 17F |
Gene Aliases: |
C87042, IL-17F |
Location: |
Chr.1:20777146-20785274 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Il17f | NM_145856.2 | 3 | 742 | NP_665855.2 |
| AK157101.1 | 1 | 411 | BAE33962.1 | |
| AY380822.1 | 3 | 742 | AAQ88439.1 |

Genomic Map