Assay Details
Target Gene Details
Entrez Gene ID: | 18159 |
Gene Name: | natriuretic peptide type C |
Gene Aliases: |
CNP, lbab |
Location: |
Chr.1:86666293-86670573 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nppc | NM_010933.5 | 3 | 623 | NP_035063.1 |
| AK020093.1 | 3 | 627 | BAB31991.1 | |
| AK036176.1 | 3 | 625 | BAC29333.1 | |
| BC111834.1 | 3 | 554 | AAI11835.1 |
More Information
Set Membership: |
|

Genomic Map