Assay Details
Target Gene Details
Entrez Gene ID: | 215378 |
Gene Name: | bone morphogenetic protein/retinoic acid inducible neural specific 3 |
Gene Aliases: |
B830045N13Rik, Fam5c |
Location: |
Chr.1:146495666-146902474 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Brinp3 | NM_001145807.1 | 8 | 2271 | NP_001139279.1 |
| NM_153539.3 | 8 | 2629 | NP_705767.3 | |
| AK046878.1 | 8 | 2630 | ||
| AK083233.1 | 9 | 2320 | ||
| BC030498.1 | 7 | 2247 | AAH30498.1 | |
| BC099956.1 | 8 | 2606 | AAH99956.1 |

Genomic Map