Assay Details
Target Gene Details
Entrez Gene ID: | 18986 |
Gene Name: | POU domain, class 2, transcription factor 1 |
Gene Aliases: |
2810482H01Rik, NF-A1, Oct-1, Oct1, Otf-1, Otf1 |
Location: |
Chr.1:165865150-166002672 on Build GRCm38 |
Assay Gene Location: | Within Exon 16 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pou2f1 | NM_011137.3 | 15 | 1766 | NP_035267.2 |
NM_198932.3 | 14 | 1694 | NP_945150.2 | |
NM_198933.3 | 14 | 1766 | NP_945151.2 | |
NM_198934.3 | 15 | 1838 | NP_945152.2 | |
AF508939.1 | 14 | 1675 | AAM34281.1 | |
AJ296212.1 | 14 | 1718 | CAC34943.1 | |
AJ489474.1 | 15 | 1838 | CAD35743.1 | |
AJ489475.1 | 13 | 1583 | CAD35744.1 | |
AK028237.1 | 15 | 1768 | BAC25832.1 | |
AK082573.1 | 2 | 428 | BAC38535.1 | |
S65461.1 | 13 | 1594 | AAB28234.1 | |
X56230.1 | 13 | 1771 | CAA39679.1 | |
X68362.1 | 15 | 1739 | CAA48422.1 | |
X68363.1 | 14 | 1665 | CAA48423.1 | |
X68364.1 | 14 | 1681 | CAA48424.1 | |
X70324.1 | 13 | 1598 | CAA49791.1 | |
X70325.1 | 4 | 507 | CAA49792.1 |
