Assay Details
Target Gene Details
Entrez Gene ID: | 320119 |
Gene Name: | ribosomal protein S6 kinase polypeptide 1 |
Gene Aliases: |
AA682037, B130003F20Rik, C80612, Rpk118 |
Location: |
Chr.1:190772879-190912139 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rps6kc1 | NM_178775.4 | 6 | 706 | NP_848890.3 |
| AK044809.1 | 3 | 425 | BAC32101.1 | |
| AK149858.1 | 6 | 671 | BAE29128.1 | |
| BB619353.1 |
More Information
Set Membership: |
|

Genomic Map