Assay Details
Target Gene Details
Entrez Gene ID: | 67223 |
Gene Name: | ribosomal RNA processing 15 homolog (S. cerevisiae) |
Gene Aliases: |
2810430M08Rik, 5430437H21Rik |
Location: |
Chr.1:186721086-186749358 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Rrp15 | NM_026041.2 | 5 | 907 | NP_080317.3 |
AK007616.1 | 5 | 861 | BAB25137.2 | |
AK010658.1 | 5 | 875 | BAB27096.1 | |
AK013204.1 | 5 | 884 | ||
AK013217.1 | 5 | 875 | BAB28719.2 | |
AK087997.1 | 5 | 877 | BAC40083.1 | |
AK169088.1 | 5 | 908 | BAE40872.1 | |
BC006787.1 | 5 | 873 | AAH06787.1 | |
CO043575.1 | 1 | 322 |
