Assay Details
Target Gene Details
Entrez Gene ID: | 23874 |
Gene Name: | phenylalanyl-tRNA synthetase, beta subunit |
Gene Aliases: |
C76708, Farsa, Farsl, Farslb, Frsb |
Location: |
Chr.1:78417958-78488897 on Build GRCm38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Farsb | NM_001278075.1 | NP_001265004.1 | ||
| NM_011811.4 | 17 | 2012 | NP_035941.2 | |
| NR_102757.1 | 17 | 1914 | ||
| NR_102758.1 | 16 | 1956 | ||
| AK012154.1 | BAB28064.1 | |||
| AK045432.1 | 17 | 1964 | BAC32363.1 | |
| AK134799.1 | 16 | 1918 | BAE22290.1 | |
| AK187456.1 | ||||
| BM212593.2 | 1 | 263 |
More Information
Set Membership: |
|

Genomic Map