Assay Details
Target Gene Details
Entrez Gene ID: | 72585 |
Gene Name: | Ly6/Plaur domain containing 1 |
Gene Aliases: |
2700050C12Rik, AI853408, C530008O16Rik, Lynx2, Lypdc1 |
Location: |
Chr.1:125867635-125913186 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lypd1 | NM_145100.3 | 1 | 322 | NP_659568.2 |
| AB041649.1 | 1 | 322 | BAA95101.1 | |
| AK045592.1 | 1 | 323 | BAC32428.1 | |
| AK082941.1 | BAE43380.1 | |||
| BC058599.1 | 1 | 255 | AAH58599.1 |

Genomic Map