Assay Details
Target Gene Details
Entrez Gene ID: | 100502590 |
Gene Name: | RIKEN cDNA 4930469G21 gene |
Gene Aliases: |
- |
Location: |
Chr.1:161156843-161159314 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 4930469G21Rik | NM_001195189.1 | NP_001182118.1 |
Target Gene Details
Entrez Gene ID: | 73844 |
Gene Name: | ankyrin repeat domain 45 |
Gene Aliases: |
4933409K03Rik |
Location: |
Chr.1:161142547-161170507 on Build GRCm38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ankrd45 | NM_028664.1 | NP_082940.1 | ||
| AK016757.1 | BAB30413.1 | |||
| BC049713.1 | AAH49713.1 |
More Information
Set Membership: |
|

Genomic Map