Assay Details
Target Gene Details
Entrez Gene ID: | 66153 |
Gene Name: | F-box protein 36 |
Gene Aliases: |
0610008D19Rik, 1110020F21Rik, 2410002G19Rik, D1Ertd757e |
Location: |
Chr.1:84839841-84900487 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fbxo36 | NM_025386.3 | 4 | 647 | NP_079662.1 |
| AK002325.1 | 4 | 627 | ||
| AK003856.1 | 4 | 647 | BAB23040.1 | |
| AK008324.1 | 4 | 644 | BAB25602.1 | |
| AK010335.1 | 4 | 1460 | ||
| AU019733.1 | 1 | 254 | ||
| BC049581.1 | 4 | 624 | AAH49581.1 | |
| BY708252.1 | 4 | 644 |

Genomic Map