Assay Details
Target Gene Details
Entrez Gene ID: | 17164 |
Gene Name: | MAP kinase-activated protein kinase 2 |
Gene Aliases: |
AA960234, MAPKAP-K2, MK-2, MK2, Rps6kc1 |
Location: |
Chr.1:131053700-131097543 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mapkapk2 | NM_008551.1 | 1 | 174 | NP_032577.1 |
| AK188700.1 | ||||
| AK204522.1 | ||||
| BC063064.1 | 1 | 174 | AAH63064.1 | |
| X76850.1 | 1 | 153 | CAA54183.1 |
More Information
Set Membership: |
|

Genomic Map